Learn More
Ismatec™ Tygon™ MHSL 2001 3-stop Tubing
Product Code. 10337902
Click to view available options
Tubing Size:
0.64mm I.D., 2.44mm O.D.
1.02mm I.D., 2.72mm O.D.
1.52mm I.D., 3.22mm O.D.
2.06mm I.D., 3.76mm O.D.
2.79., I.D., 4.49mm O.D.
Unit Size:
Pack of 6
Product Code. 10337902
Brand: Ismatec™ SC0806
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Transparent, plasticizer-free 3-stop Tygon™ tubing with superior pump-life. Ismatec™ Tygon™ MHSL 2001 2-stop Tubing is especially designed for MEK and other aggressive solvents.
  • Plasticizer and oil-free.
  • Smooth inner-bore.
  • Low sorption maintains fluid and tube integrity.
  • Does not impart anything into the pumping medium.
  • No release of hazardous materials when properly incinerated.
  • Applications
    • Acids: Excellent
    • Alkaline solutions: Excellent
    • Solvents: Good/Excellent
  • Complies with: FDA.
  • Sterilization
    • Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 121°C.
    • Gas sterilization with ethylene oxide.
    • Sterilization with radiation up to 2.5 mrad.
  • Note: Although designed for MS/CA cassettes, this tubing has only 2 stoppers!
Specifications
Certifications/Compliance FDA
Description Color-code: White-White
Autoclavable Autoclavable
Material Tygon MHSL 2001
Overall Length (Metric) 30 cm
Thickness (Metric) 0.85 mm
For Use With (Application) MEK and other aggressive solvents. MS/CA cassettes.
Min. Operating Temperature (Metric) -73°C
Packaging Quantity 6 pcs./Pk.
Tubing Size 1.02mm I.D., 2.72mm O.D.
Gas Permeability CO2 1140, O2 76, N2 190
Color Transparent
Max. Operating Temperature (Metric) 57°C
Product Type 3-stop Tubing