Learn More
Fisherbrand™ Sterile Polyethylene Sampling Bags
Designed with a twirl round metal wire closure system and puncture proof safety tabs. Fisherbrand™ Sterile Polyethylene Sampling Bags are leakproof when sealed.
Brand:  Fisherbrand™ EPR-3050-FS-1_OLD
Product Code. 11502783
  • € 173.33 / Pack of 1000
Estimated Shipment: 23-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured with pliant, highly resistant virgin polyethylene 
  • Sterile transparent polyethylene bags without side seals
  • Choose clear or white marking area for written notations
  • Convenient pull tabs to open bag without touching the edges
  • Ideal for use with solid, semi-solid or liquid samples
  • For use in all laboratories where samples need to be collected into a sterile environment.
Specifications
Specifications
60 mL
Use in all laboratories where samples need to be collected into a sterile environment
Sterile
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Sterile Polyethylene Sampling Bags > Capacity: 60mL