Learn More
Fisherbrand Twirl'EM™ Clear Polyethylene Sampling Bags
Click to view available options
Capacity (Metric):
150 mL
650 mL
800 mL
1060 mL
1240 mL
2250 mL
4000 mL
Unit Size:
Pack of 1000
Specifications
Specifications
| Capacity (Metric) | 1240 mL |
| Color | Clear |
| Material | Polyethylene |
| Sterility | Sterile |
| Quantity | 1000 Pk. |