Learn More
Thermo Scientific™ Tube Rocker Stacking Bars
Accessory for use with Digital Tube Rocker, 120V and Digital Tube Rocker, 230V
Brand: Thermo Scientific™ 88882160
48.83 EUR valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 0.20000kg
Description
Double your capacity in a compact footprint while using the Thermo Scientific™ Digital Tube Rocker with the Tube Rocker Stacking Bars.
Specifications
Stainless Steel | |
Tube Rocker Stacking Bars | |
Digital Tube Rocker, 120V (88882017), Digital Tube Rocker 230V (88882018) |
0.8 x 6 cm | |
Platform Tray (88882158), Tube Rocker Stacking Bars (88882160), Variety of tubes | |
4 stacking bars for 2nd tube rocker tray installation |