Learn More
Gosselin™ Tube Closures
Suitable for different usage of tubes and applications. Gosselin™ Tube Closures are offered in straight, winged, skirted, or screw cap styles and are manufactured from PE plastic.
Brand:  Gosselin™ B135-23
 View more versions of this product
Product Code. 11922815
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • All closures vertically ribbed for easier grip
  • Protected with a polyethylene outer bag to prevent particle contamination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EC N°1935/2004, ISO 11137
LDPE
13.5 mm Tube
Straight Plug
White
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gosselin™ Tube Closures > Straight Cap; Fits 13.5mm tube; White