Learn More
Multiroir Controlec™ Trolley
Click to view available options
Dimensions (L x W x H):
1100 x 520 x 1020 mm (Overall), 504 x 910 mm (Tray)
1100 x 520 x 950 mm (Overall), 504 x 910 mm (Tray)
480 x 800 x 1000 mm (Overall), 435 x 615 mm (Tray)
480 x 800 x 950 mm (Overall), 435 x 615 mm (Tray)
Color:
Black
Light grey
No. of Shelves:
2
3
Unit Size:
Each
Description
- Aluminum frame with handles at each end
- Available in two or three tray format
- On casters for easy manouvrability
- Available in light grey or black
Specifications
Specifications
| Color | Black |
| Dimensions (L x W x H) | 480 x 800 x 1000 mm (Overall), 435 x 615 mm (Tray) |
| Load Bearing Capacity (Metric) | 180 kg (Overall) |
| Material | Aluminum Frame, Polypropylene Trays |
| No. of Shelves | 3 |
| Shelf Material | Polypropylene |
| Wheel Type | Lockable casters |