missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals TRIS-glycine-native running buffer (10X), pH 8.5
POA - € 79.50
Specifications
Chemical Name or Material | TRIS-glycine-native running buffer (10X) |
---|---|
Physical Form | Liquid |
Packaging | Plastic bottle |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15425209
|
Thermo Scientific Chemicals
J62914.K2 |
1 L |
€ 48.74
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15435209
|
Thermo Scientific Chemicals
J62914.K3 |
2 L |
€ 79.50
2 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15445209
|
Thermo Scientific Chemicals
J62914.K7 |
4 L | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
TRIS-glycine-native running buffer (10X) | |
Liquid | |
Plastic bottle |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only