missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris-Borate-Edta Buffer, for Electrophoresis, (10X Concentration), Fisher Chemical™
Click to view available options
Quantity:
1 L
10 L
Packaging:
Amber glass bottle
Bag in box
Description
Tris-Borate-Edta Buffer, for Electrophoresis, (10X Concentration)
Specifications
Specifications
Boiling Point | 219°C |
Color | Colorless |
pH | 8.2 to 8.4 |
Physical Form | Liquid |
Quantity | 1 L |
Packaging | Amber glass bottle |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'