missing translation for 'onlineSavingsMsg'
Learn More

Triisopropyl phosphite, 90+%, Thermo Scientific Chemicals

Product Code. 11433910
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11433910

Brand: Thermo Scientific Alfa Aesar A15247.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-17-6
Molecular Formula C9H21O3P
Molecular Weight (g/mol) 208.238
MDL Number MFCD00008874
InChI Key SJHCUXCOGGKFAI-UHFFFAOYSA-N
Synonym triisopropyl phosphite, tri-2-propyl phosphite, phosphorous acid, tris 1-methylethyl ester, isopropyl phosphite, tri, triisopropoxyphosphine, phosphorous acid, triisopropyl ester, tri-2-propylphosphite, unii-rj7qu4yqun, triisopropylphosphite, tri-i-propylphosphite
PubChem CID 8304
IUPAC Name tripropan-2-yl phosphite
SMILES CC(C)OP(OC(C)C)OC(C)C

Specifications

Density 0.908
Boiling Point 63°C to 64°C (11 mmHg)
Flash Point 46°C (114°F)
Odor Characteristic
Refractive Index 1.411
Quantity 500 mL
UN Number UN1993
Beilstein 1701528
Sensitivity Air and moisture sensitive
Formula Weight 208.24
Percent Purity ≥90%
Chemical Name or Material Triisopropyl phosphite
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.