missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals
Shop All Thermo Scientific Products
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent

Specifications
Specifications
Melting Point | -63.5°C |
Density | 1.5100g/mL |
Boiling Point | 39.1°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (GC) |
Packaging | Glass bottle |
Linear Formula | (CF3CO)2O |
Refractive Index | 1.3 |
Quantity | 250 g |
Beilstein | 02, II, 186 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0452441 | Certificate of Analysis | 23/04/2024 | 147810025,147812501,147811000,147810250,147815000 |
A0447418 | Certificate of Analysis | 09/11/2023 | 147810025,147812500,147812501,147811000,147810250,147815000 |
A0342245 | Certificate of Analysis | 12/12/2022 | 147810025,147812500,147812501,147811000,147810250,147815000 |
A0344679 | Certificate of Analysis | 12/12/2022 | 147810025,147812500,147812501,147811000,147810250,147815000 |
A0354408 | Certificate of Analysis | 12/12/2022 | 147810025,147812500,147812501,147811000,147810250,147815000 |
Safety and Handling


Product Identifier
- Trifluoroacetic anhydride
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- EUH014-Reacts violently with water.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Trifluoroacetic acidanhydride
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction