missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyltin chloride, 95%, tech.

Product Code. p-202280 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10755811

Brand: Thermo Scientific Acros 139351000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1461-22-9 | C12H27ClSn | 325.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1461-22-9
Molecular Formula C12H27ClSn
Molecular Weight (g/mol) 325.48
MDL Number MFCD00000521
InChI Key GCTFWCDSFPMHHS-UHFFFAOYSA-M
Synonym tributyltin chloride, chlorotributyltin, tri-n-butyltin chloride, tributylchlorotin, stannane, tributylchloro, chlorotri-n-butyltin, tri-n-butylchlorotin, tributyl chloro stannane, chlorotributylstannane, tributylstannyl chloride
PubChem CID 15096
ChEBI CHEBI:79734
IUPAC Name tributyl(chloro)stannane
SMILES CCCC[Sn](CCCC)(CCCC)Cl

Specifications

Melting Point -9.0°C
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 171.0°C to 173.0°C (25.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Linear Formula [CH3(CH2)3]3SnCl
Refractive Index 1.4880 to 1.4940
Quantity 100 g
Merck Index 15, 9782
Specific Gravity 1.2
Solubility Information Solubility in water: 0.07g/L (20°C). Other solubilities: soluble in most common orgnaic solvents,including,benzene and toluene
Viscosity 7 mPa.s (20°C)
Formula Weight 325.48
Percent Purity 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material Tri-n-butyltin chloride, tech. 90%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tri-n-butyltin chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360FD-May damage fertility. May damage the unborn child.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.