Learn More
Deutsch Neumann™ Silicone Rubber Stoppers
Click to view available options
Diameter (Metric) Bottom:
5 mm
8 mm
12.5 mm
17 mm
23 mm
31 mm
36 mm
Unit Size:
Each
Description
- Acc. to DIN 12 871
- Heat resistant up to 200°C
- Durometer 50 Shore A
- Available in a wide range of sizes
Specifications
Specifications
| Height (Metric) | 40mm |
| Quantity | 1/Ea. |
| Color | Transparent |
| Diameter (Metric) Bottom | 36 mm |
| Diameter (Metric) Top | 44 mm |
| Material | Silicone |
| Product Type | Silicone Stopper |