Learn More
Deutsch Neumann™ Silicone Tubing
Click to view available options
Diameter (Metric) Inner:
0.5 mm
1 mm
1.5 mm
2 mm
3 mm
4 mm
5 mm
7 mm
8 mm
9 mm
10 mm
13 mm
14 mm
15 mm
16 mm
18 mm
20 mm
Diameter (Metric) Outer:
1.3 mm
1.8 mm
2.3 mm
3 mm
3.5 mm
3.8 mm
6 mm
7 mm
8 mm
12 mm
13 mm
14 mm
15 mm
16 mm
17 mm
18 mm
19 mm
20 mm
21 mm
24 mm
26 mm
Unit Size:
15m
1m
25m
5m
Each
Description
- Temperature range of -60°C to +200°C (short periods up to +260°C)
- Meets following standards: BFR XV, FDA CFR § 177.2600, EP - Europ. Pharmacopoeia
- Test for biocompatibility according to ISO 10993-1
- Durometer 60 Shore A
Specifications
Specifications
| Certifications/Compliance | BFR XV, FDA CFR 21§ 177.2600, Europ. Pharmacopoeia Test, ISO 10993-1 |
| Diameter (Metric) Inner | 9 mm |
| Material | Silicone |
| Diameter (Metric) Outer | 12 mm |
| Overall Length (Metric) | 25 m |
| Thickness (Metric) | 1.5 mm |
| Color | Transparent |