Learn More
VITLAB™ Transparent Polypropylene Watch Glasses
€ 41.65 - € 63.55
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
15292749
|
VITLAB™
80454 |
€ 41.65
Pack of 10 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||||
15202759
|
VITLAB™
80456 |
118.5 mm |
€ 63.55
Pack of 10 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||||
10748837
|
VITLAB™
80455 |
100 mm |
€ 55.70
Pack of 10 |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
Specifications
Polypropylene |