Learn More
Vitlab™ Transparent HDPE Large Funnel
Click to view available options
Diameter (Metric)Top:
200 mm
Unit Size:
Pack of 6
Description
- Transparent
- Rapid flow due to 60° angle
- Practical handle for hanging
- Optional accessories available
Specifications
Specifications
| Diameter (Metric)Top | 200 mm |
| Capacity (Metric) | 1300 mL |
| Material | HDPE |
| Height (Metric) | 200 mm |
| Description | HDPE filter |
| For Use With (Application) | Suitable for filling large amounts of liquids |