Learn More
Fisherbrand™ Translucent Silicone Stoppers
Prevents liquid chemical leaks or escape from container.
€ 10.00 - € 71.00
Specifications
Color Translucent
Material Silicone
Product Type Silicone Stopper
View More Specs
Products 23
Product Code Brand Diameter (Metric) Bottom Price Quantity & Availability  
Product Code Brand Diameter (Metric) Bottom Price Quantity & Availability  
11507662
View Documents
Fisherbrand™
SB010/S
10 mm
€ 16.00
Pack of 20
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
11517662
View Documents
Fisherbrand™
SB011/S
11 mm
€ 11.00
Pack of 10
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
11537662
View Documents
Fisherbrand™
SB015/S
15 mm
€ 19.00
Pack of 10
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
11557662
View Documents
Fisherbrand™
SB019/S
19 mm
€ 17.00
Pack of 5
Estimated Shipment: 10-01-2025
to see stock.
Add to basket
11567662
View Documents
Fisherbrand™
SB021/S
21 mm
€ 19.00
Pack of 5
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
11587662
View Documents
Fisherbrand™
SB025/S
25 mm
€ 10.00
Pack of 2
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
11577662
View Documents
Fisherbrand™
SB023/S
23 mm
€ 23.00
Pack of 5
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
11597662
View Documents
Fisherbrand™
SB027/S
27 mm
€ 14.00
Pack of 2
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
11507672
View Documents
Fisherbrand™
SB029/S
29 mm
€ 15.00
Pack of 2
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
11527672
View Documents
Fisherbrand™
SB033/S
33 mm
€ 24.00
Pack of 2
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
11537672
View Documents
Fisherbrand™
SB035/S
35 mm
€ 15.00
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
11547672
View Documents
Fisherbrand™
SB037/S
37 mm
€ 15.00
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
11557672
View Documents
Fisherbrand™
SB039/S
39 mm
€ 17.00
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
11567672
View Documents
Fisherbrand™
SB041/S
41 mm
€ 21.00
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
11587672
View Documents
Fisherbrand™
SB045/S
45 mm
€ 25.00
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
11547652
missing translation for 'viewDocuments'
Fisherbrand™
SB004/S
4 mm
€ 14.00
Pack of 50
Estimated Shipment: 15-01-2025
to see stock.
Añadir a la cesta
11557652
missing translation for 'viewDocuments'
Fisherbrand™
SB005/S
5 mm
€ 15.00
Pack of 50
Estimated Shipment: 14-01-2025
to see stock.
Añadir a la cesta
11517682
missing translation for 'viewDocuments'
Fisherbrand™
SB057/S
57 mm
€ 43.00
Each
Estimated Shipment: 05-02-2025
to see stock.
Añadir a la cesta
11527682
missing translation for 'viewDocuments'
Fisherbrand™
SB062/S
62 mm
€ 57.00
Each
Estimated Shipment: 30-01-2025
to see stock.
Añadir a la cesta
11537682
missing translation for 'viewDocuments'
Fisherbrand™
SB067/S
67 mm
€ 64.00
Each
Estimated Shipment: 14-01-2025
to see stock.
Añadir a la cesta
11547682
missing translation for 'viewDocuments'
Fisherbrand™
SB071/S
71 mm
€ 71.00
Each
Estimated Shipment: 31-01-2025
to see stock.
Añadir a la cesta
11577652
View Documents
Fisherbrand™
SB007/S
7 mm
€ 19.00
Pack of 50
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
11597652
View Documents
Fisherbrand™
SB009/S
9 mm
€ 13.00
Pack of 20
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
Description
Description
  • Non-toxic, translucent
  • Temperature range -60°C to 200°C
  • High electrical resistance
  • High resistance to many chemicals including ozone
Specifications
Specifications
Translucent
Silicone
Silicone Stopper