Learn More
Fisherbrand™ Translucent Platinum-Cured Silicone Tubing
Product Code. 11758168 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Inner:
0.5 mm
0.8 mm
1.6 mm
2.4 mm
3.2 mm
4.8 mm
6.4 mm
12.8 mm
16 mm
Diameter (Metric) Outer:
2.1 mm
2.4 mm
3.2 mm
4 mm
4.8 mm
6.4 mm
8 mm
9.6 mm
16 mm
22.4 mm
Overall Length (Metric):
25 m
5 m
Unit Size:
25m
5m
Each
Product Code. 11758168
Only null left
Add to basket
Add to basket
Use this tubing for pharmaceutical, biotech, peristaltic pumping and food applications. Fisherbrand™ Translucent Platinum-Cured Silicone Tubing is odorless, nontoxic and features excellent heat resistance.
  • Translucent
  • Non-yellowing
  • No peroxide by-products
  • Standard high tear strength
  • High gloss finish
  • Excellent quality
  • Highly hydrophobic
  • Extended life in Peristaltic pumps
Specifications
Certifications/Compliance FDA, BFR and WRAS approved
Autoclavable Autoclavable
Diameter (Metric) Inner 12.8 mm
Material Platinum-cured Silicone
Diameter (Metric) Outer 16 mm
Overall Length (Metric) 5 m
Thickness (Metric) 1.6 mm
For Use With (Application) Biological , pharmaceutical, biotechnology, aerospace, hot work
Min. Operating Temperature (Metric) -40°C
Sterility Sterile
Color Translucent
Flexibility Flexible
Max. Operating Temperature (Metric) 200°C