missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxy-D-proline, 97%

Product Code. p-7029902
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15423687

Brand: Thermo Scientific Alfa Aesar H27421.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3398-22-9 | C5H9NO3 | 131.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3398-22-9
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00065608
InChI Key PMMYEEVYMWASQN-UHFFFAOYNA-N
Synonym trans-4-hydroxy-d-proline, 2r,4s-4-hydroxypyrrolidine-2-carboxylic acid, h-d-hyp-oh, d-proline, 4-hydroxy-, 4s, 2r,4s-4-hydroxy-pyrrolidine-2-carboxylic acid, d-hydroxyproline, ambotzhaa1169, pubchem9966, d-hyp-oh
PubChem CID 440074
ChEBI CHEBI:27992
IUPAC Name (2R,4S)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES OC1CNC(C1)C(O)=O

Specifications

Melting Point ∽260°C (decomposition)
Quantity 250 mg
Formula Weight 131.13
Percent Purity 97%
Chemical Name or Material trans-4-Hydroxy-D-proline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.