Learn More
Thermo Scientific™ TraceGOLD TG-VRX GC Columns
Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
105 m
20 m
30 m
40 m
60 m
75 m
Diameter (Metric):
0.18 mm
0.25 mm
0.32 mm
0.53 mm
Unit Size:
Each
Description
Thermo Scientific™ TraceGOLD TG-VRX GC Columns provide good resolution and fast analysis times for volatile compounds.
- Proprietary stationary phase
- Optimized dimensions for analysis of volatile organic pollutants
- Ideally suited for US EPA Methods 502.2, 524.2, 8021, and 8260
- Similar to DB-VRX, Rtx-VRX
Recommended for:
- Volatile organic pollutants
- Aromatics
Specifications
Specifications
| Max. Temperature | 260°C |
| Diameter (Metric) | 0.53 mm |
| Diameter (Metric) Inner | 0.53 mm |
| Stationary Phase | TraceGOLD™ TG-VRX |
| For Use With (Application) | GC/MS for Volatile Organic Pollutants, GC/MS for US EPA Method 8021 Compounds |
| Film Thickness | 3 μm |
| Length (Metric) | 75 m |
| Polarity | Non-Polar |
| Description | 75m x 0.53mm x 3.00um |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur