Learn More
Thermo Scientific™ TraceGOLD™ TG-5MT Metal GC Columns
Shop All Thermo Scientific ProductsDescription
TG-5MT Metal GC Columns provide greater robustness under increased stress.
- Can be used at elevated temperature without risk of bleed or breakdown
- Excellent inertness when working with highly active compounds
- Robust under stress
Specifications
Specifications
| Max. Temperature | 330/350°C (Standard Columns), 430°C (Metal Columns) |
| Diameter (Metric) | 0.53 mm |
| Diameter (Metric) Inner | 0.53 mm |
| Stationary Phase | TraceGOLD™ TG-5 MT |
| USP Type | G27, G36 |
| For Use With (Application) | Solvent Impurities Analysis, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes |
| Particle Size | 1.5 μm |
| Temperature Range | -60°C to +430°C |
| Film Thickness | 1.5 μm |
| Length (Metric) | 30 m |
| Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.