Learn More
Hach Lange™ Total Chlorine Reagent Powder Pillows
Product Code. 12046802
Click to view available options
Volume (Metric):
5 mL
10 mL
Unit Size:
Each
Product Code. 12046802
Brand: Hach Lange™ 1407699
Only null left
Add to basket
Add to basket
For total chlorine determination by the DPD method.
For use with Chlorine Visual Test Kits up to 3.5 mg/L Cl2.
    • Total Chlorine Reagent Powder Pillows, DPD (pk/100)
    • For use with Chlorine Visual Test Kits up to 3.5 mg/L Cl2
    • For 5 mL samples
    • Features Chlorine, Bromine, Iodine parameters
Specifications
Volume (Metric) 5 mL
Format Powder
For Use With (Application) Hach's Chlorine Visual Test Kits
For Use With (Equipment) Hach's Chlorine Visual Test Kits
Parameter Specifications Chlorine, Total
Quantity 100 Pack
Range 0 to 3.5 mg/L
Test Method DPD
Type Total Chlorine Reagent Powder Pillows
compliance-icons
Product Identifier
  • DPD Total Chlorine Reagent
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : di-Sodium hydrogen phosphate, Potassiumiodide, Salt of N, N-Diethyl-p-Phenylenediamine