missing translation for 'onlineSavingsMsg'
Learn More

SCA Hygiene Products™ Tork™ Wiping Papers

Brand:  SCA Hygiene Products™ 130109

 View more versions of this product

Product Code. 10083574

  • € 219.00 / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Wiping Paper
34 cm
Food contact approved
1180m x 34 cm
Multipurpose - handles most wiping tasks, Ideal for glass cleaning
1180 m
Paper
White
Roll
1-ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers