Learn More
SCA Hygiene Products™ Tork Virgin Fibre Wiping Paper Plus
Brand: SCA Hygiene Products™ 101221_CLEAN
This item is not returnable.
View return policy
Specifications
Wiping Paper Plus | |
215 mm | |
ISO 9001, ISO 14001 | |
Single-use | |
Mini Centre feed Roll | |
11/Roll |
75 m | |
Virgin Fiber | |
White | |
75 m x 215 mm | |
Tork Portable Mini Centre feed Disp Accessory, Tork Centre feed Dispenser, Tork Centre feed Starter Pack, Tork Metal Centre feed Dispenser | |
2-ply |