missing translation for 'onlineSavingsMsg'
Learn More

SCA Hygiene Products™ Tork™ Basic Papers

Brand:  SCA Hygiene Products™ 129262_CLEAN

 View more versions of this product

Product Code. 10276914

  • € 122.00 / Pack of 2
Estimated Shipment: 26-11-2024
to see stock.
Explore more special offers
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Description

Description

Specifications

Specifications

Universal Wipers
24 cm
Food contact approved
340m x 23.5 cm
Use with System W1 Tork floor, wall or W2 combi dispensers for improved hygiene and convenience
340 m
Paper
White
Combi Roll
2-ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers