missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Toluidine Blue O, pure

Product Code. 10135113 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10135113

Brand: Thermo Scientific Chemicals 348601000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 92-31-9 | C15H16ClN3S | 305.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-31-9
Molecular Formula C15H16ClN3S
Molecular Weight (g/mol) 305.82
MDL Number MFCD00011934
InChI Key HNONEKILPDHFOL-UHFFFAOYSA-M
Synonym Basic blue 17, C.I. 52040, Tolonium chloride
PubChem CID 7083
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1

Specifications

Beilstein 27, 402
Merck Index 15, 9679
Solubility Information Solubility in water: 38.2g/L. Other solubilities: soluble in alcohol
Infrared Spectrum Authentic
IUPAC Name (7-amino-8-methylphenothiazin-3-ylidene)-dimethylazanium;chloride
Formula Weight 305.82
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Green
Quantity 100 g
Physical Form Powder
Chemical Name or Material Toluidine blue O
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.