missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, Puriss. p.a., ACS Reagent, ≥99.7% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
25 mL
50 mL
4 x 25 mL
5 L
10 L
Packaging:
Aluminum Bottle
Glass Bottle
Stainless Steel Can
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
Melting Point | -95°C |
Color | Colorless |
Density | 0.865g/mL at 25°C |
Boiling Point | 110.6°C |
Quantity | 10 L |
Explosion Limit | 0.07 |
Flash Point | 4°C (39.2°F) |
Ignition Point | 997°F |
Odor | Sweet pungent |
Packaging | Stainless Steel Can |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'