Learn More
Tocris Bioscience™ PACAP 1-38
Potent stimulator of adenylyl cyclase
Brand:  Tocris Bioscience™ 1186/100U
223.10 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18085522
  • € 285.00 / 100µg
Explore more special offers
Description
Description
PACAP 1-38 is a potent endogenous neuropeptide (IC50 = 2 nM) showing considerable homology with vasoactive intestinal peptide (VIP) but with a greater potency for stimulation of adenylyl cyclase. PACAP 1-38 induces phosphorylation of NR2B and enhances NMDA receptor potentials.
Specifications
Specifications
95%
Store at -20°C
100 μg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers