Learn More
Tocris Bioscience™ PACAP 1-38
Potent stimulator of adenylyl cyclase
Brand: Tocris Bioscience™ 1186/100U
223.10 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
PACAP 1-38 is a potent endogenous neuropeptide (IC50 = 2 nM) showing considerable homology with vasoactive intestinal peptide (VIP) but with a greater potency for stimulation of adenylyl cyclase. PACAP 1-38 induces phosphorylation of NR2B and enhances NMDA receptor potentials.Specifications
95% | |
Store at -20°C | |
100 μg |