Learn More
Tocris Bioscience™ Orexin B (mouse)
Endogenous agonist at OX1 and OX2
Brand: Tocris Bioscience™ 1457/500U
256.74 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Orexin B (mouse) is an endogenous agonist at orexin receptors. May be involved in the regulation of feeding, sleep-wake cycle and other hypothalamic functions. Orexin B exerts excitatory effects on nigral dopaminergic neurons and alleviates motor symptoms in a Parkinson's disease mouse model.Specifications
95% | |
Store at -20°C | |
500 μg |