Learn More
Tocris Bioscience™ NFAT Inhibitor
Inhibitor of calcineurin-mediated NFAT activation
Brand:  Tocris Bioscience™ 3930/1
Product Code. 18064032
  • € 304.00 / 1mg
Explore more special offers
Description
Description
NFAT Inhibitor is a selective inhibitor of calcineurin-mediated dephosphorylation of nuclear factor of activated T cells (NFAT). Does not disrupt other calcineurin-dependent pathways. Inhibits NFAT activation and NFAT-dependent expression of endogenous cytokine genes in T cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.95