Learn More
Tocris Bioscience™ [D-p-Cl-Phe6,Leu17]-VIP
Selective VIP receptor antagonist
Brand:  Tocris Bioscience™ 3054/1
313.41 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18095082
  • € 387.00 / 1mg
Explore more special offers
Description
Description
[D-p-Cl-Phe6,Leu17]-VIP is a selective vasoactive intestinal peptide (VIP) receptor antagonist (IC50 = 125.8 nM). Displays no activity on glucagon, secretin or GRF receptors.
Specifications
Specifications
1 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers