Learn More
Tocris Bioscience™ Astressin 2B
Selective CRF2 antagonist
Brand: Tocris Bioscience™ 2391/500U
458.60 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Astressin 2B is a potent and selective corticotropin-releasing factor receptor 2 (CRF2) antagonist (IC50 values are 1.3 and > 500 nM for CRF2 and CRF1 respectively). Antagonizes CRF2-mediated inhibition of gastric emptying.Specifications
500 μg |