Learn More
TNF-alpha Antibody (6N1E7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-27223
 View more versions of this product
Product Code. 18188632
  • € 488.00 / 0.10mg
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Inhibition Activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TNF-alpha
Monoclonal
0.5 mg/mL
Western Blot 2 - 4 ug/mL, Flow Cytometry 0.5 - 2 ug/10^6 cells, Inhibition Activity
APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Mouse
Protein G purified
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, Inhibition Assays
6N1E7
Unconjugated
P01375
TNF
Full-length recombinant human TNF alpha was used as the immunogen for this antibody.
0.1 mg
Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
7124
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1 κ