missing translation for 'onlineSavingsMsg'
Learn More

Titanium powder, -100 mesh, 99.4%

Product Code. p-7048391
Click to view available options
Quantity:
25 g
500 g
Unit Size:
25g
500g
This item is not returnable. View return policy

Product Code. 15460987

Brand: Thermo Scientific Alfa Aesar 010383.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-32-6 | Ti | 47.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Color Gray
Quantity 25 g
Mesh Size −100 Mesh
Odor Odorless
Form Powder
UN Number UN2546
Percent Purity 99.4%
Chemical Name or Material Titanium powder
compliance-icons
Product Identifier
  • Titanium powder
Signal Word
  • Danger
Hazard Category
  • Pyrophoric solid Category 1
  • Self-heating substance Category 2
Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H252-Self-heating in large quantities; may catch fire.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use to extinguish.
  • P422-Store contents under .
  • P420-Store away from other materials.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.