missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bode Chemie™ Tissues
Brand: Bode Chemie™ 981311
This item is not returnable.
View return policy
Description
Order Info
DimensionsLxW
Specifications
Tissues | |
180 x 200 mm | |
Work surfaces, toilet seats, door handles, bedframes, and tables. |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Flammable liquid Category 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bode Chemie™ Tissues > Dimensions (L x W): 180 x 200mm
Spot an opportunity for improvement?Share a Content Correction