Learn More
Tin Standard Metal Solution 1000ppm, for AAS, Solution HCl 1M, Fisher Chemical™
Click to view available options
Quantity:
100 mL
500 mL
Specifications
Specifications
Packaging | HDPE plastic bottle |
Quantity | 100 mL |
Color | Colorless |
Physical Form | Liquid |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Tin Standard Metal Solution 1,000ppm
- Warning
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- P302+P352-IF ON SKIN: Wash with plenty of water/A§.