Learn More
Bandelin Electronic™ Tickopur R 3 Special Acid Cleaners
Brand: Bandelin Electronic™ 923
Specifications
Special Acid Cleaner | |
3 | |
Colorless | |
For cleaning, rust removal, decalcification and degreasing of disassembled and undivided Machinery, tools, equipment | |
2 L |
Liquid | |
Tickopur | |
Polyethylene Henkel bottle | |
1 Ea. |