Learn More
Codan™ Three-part Syringes
Product Code. 11950735
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
20 mL
60 mL
100 mL
Connection Type:
Catheter
Catheter Cone Luer
Centric Luer Cone
Eccentric Luer Cone
Luer Tip
Unit Size:
Pack of 100
Pack of 15
Pack of 25
Pack of 50
Pack of 80
Product Code. 11950735
Brand: Codan™ 62.8436
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use Codan™ Three-part Syringes to accommodate a variety of medical needs including: catheterization, hypodermic ear treatments, and more. The syringes are latex-free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spill
  • Sterile
  • Graduations
  • Disposable
  • Latex-free
Specifications
Autoclavable Autoclavable
Material Polypropylene
Description Dual scale; Silicone seal
Product Type Hypodermic
Disposable Single-use
Graduations 1 mL
Volume (Metric) 60 mL
Needle Point Style Catheter
Connection Type Catheter Cone Luer
Graduated Yes
Sterility Sterile