Learn More
Corning™ Thread Adapters
Shop All Corning Products
Click to view available options
Product Type:
A32/A25
A32/A28
A32/A38
A32/A40
A32/A45
A45/A32
Unit Size:
Each
Description
- Threaded
- Accessory for Bottle Top Dispenser
Specifications
Specifications
| Product Type | A32/A40 |
| For Use With (Application) | Quick and precise dispensing of acids, bases and solvents |
| For Use With (Equipment) | Bottle Top Dispenser |