Learn More
SCAT™ Polypropylene Thread Adapters
Shop All SCAT Europe Products
Click to view available options
Closure Size:
Thread 1: GL/S40 (f), Thread 2: GL45 (m)
Thread 1: S 60/61 (f) Thread 2: GL45 (m)
Thread 1: S51 (f), Thread 2: GL45 (m)
Thread 1: S55 (f), Thread 2: GL45 (m)
Thread 1: S60/61, Thread 2: S55 (m)
Closure Color:
Colorless
Unit Size:
Each
Description
SCAT™ thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
- Made of polypropylene (PP) material
- Available in a wide range of thread sizes
- Suitable for all SCAT™ caps, funnels or canisters
Specifications
Specifications
| Closure Size | Thread 1: S60/61, Thread 2: S55 (m) |
| Closure Color | Colorless |
| Liner Material | Polypropylene |
| Product Type | Thread Adapter |