Learn More
SymLine™ Thread Adapter, Type 8
Product Code. 15225144
Click to view available options
Closure Size:
GL25
Unit Size:
Each
Product Code. 15225144
Brand: SymLine™ 106417
Only null left
Add to basket
Add to basket
Dovetail your SymLine™ System to suit your exact needs. Here, you will find many useful ancillaries.
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
  • Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
  • All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Closure Size GL25
Closure Color Black
Description GL25 (m) to GL25 (m)
Color Black
Liner Material HDPE-EC
Material HDPE-EC
Product Type Thread Adapter