Learn More
Thermo Scientific Chemicals Thin-Bore Sample Tube, 2mm ID
For use with MKII Magnetic Susceptibility Balances
Brand: Thermo Scientific Chemicals 014350.KG
78.90 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- I.D.: 2mm
Specifications
1 Each | |
Thin-bore Sample Tube |
Product Suggestions
Customers who viewed this item also viewed.