Learn More
Sonoco™ ThermoSafe PolarPack™ Foam Bricks
Foam brick ice packs never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control. Refrigerant bricks are vacuum-sealed for consistent weight and shape.
Brand: Sonoco™ FPP18
36.96 EUR valid until 2026-01-16
BEST PRICE on Promo! Use promo code "24103" to get your promotional price.
Description
Refrigerant bricks are vacuum-sealed for consistent weight and shape. These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.
Specifications
| 28 Cs. | |
| 8.25 in. | |
| 20.9 cm | |
| Refrigerant Ice Pack | |
| 18 oz. | |
| 8.25 x 10 x 0.75 in. (20.9 x 20.3 x 1.9 cm) |
| 14 cm | |
| 5.5 in. | |
| 0.75 in. | |
| 1.9 cm | |
| 0.72 kg | |
| 0°C (32°F) |