Learn More
Fisherbrand™ Thermographic Measurements Temperature Strip
Product Code. 10293171 Shop All Fisherbrand Products
Encompass
Click to view available options
Temperature Range (Metric):
121°C to 160°C
77°C to 116°C
Unit Size:
Pack of 10
Product Code. 10293171
This item requires a price update prior to being ordered.
Request a Price for this item here.
The standard 8 level range covers 40°C to 260°C for general purpose temperature monitoring.
Specifications
For Use With (Application) For general purpose temperature monitoring
Product Type Temperature Strip
Max. Temperature (Metric) 116°C
Min. Temperature (Metric) 77°C
Temperature Range (Metric) 77°C to 116°C