Learn More
Thermo Scientific™ ThermoFlex Chiller
Shop All Thermo Scientific Products
Click to view available options
Cooling Capacity:
2.2 kW (20°C setpoint and 25°C ambient)
Unit Size:
Each
Description
- Internal pressure relief valve
- Custom temperature and pressure alarms
- An intuitive user interface for ease of operation
- Integrated funnel for easy filling
- Full flow fluid filter ensures clean fluid to protect your application and maximise recirculation system life
- Easily removable condenser grill and air filter allow for quick and simple cleaning to optimise chiller performance and maximise component life
- Air and water filters that can be changed while unit is in operation
- Innovative, patented packaging for rapid installation
- Quick start guide for seamless start-up in minutes
- Advanced design allows two sides of the unit to be blocked, allowing placement in a corner while maintaining full refrigeration performance
Specifications
Specifications
| Cooling Capacity | 2.2 kW (20°C setpoint and 25°C ambient) |
| Temperature Range | 5°C to 40°C |
| Type | Suitable for a range of tap water cooling applications |
| Temperature Stability | ±0.1°C accuracy |
| Electrical Requirements | 230 V 50 Hz |