Learn More
Thermo Scientific™ Sorvall™ Racks for Drum Rotor
Shop All Thermo Scientific Products
Click to view available options
Color:
Red
Tube Size:
1.5/2mL
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | Drum rotor |
| Color | Red |
| No. of Places | 10 |
| Tube Size | 1.5/2mL |