Learn More
Epredia™ Cassette 1 Biopsy Cassettes
Product Code. 15584815 Shop All Epredia Products
Click to view available options
Color:
Pink
White
Unit Size:
Pack of 500
Product Code. 15584815
Brand: Epredia™ 1000968
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specially designed to hold biopsy specimens during processing and embedding
  • Made of acetal polymer
  • Cassette writing face is at a 30° angle
  • Pore style is small square
  • Locking lid attached by two hinges
  • Supplied in a box of 500 cassettes with attached breakaway lids
Order Info
Not available in Tube Packs
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Square
Writing Face Angle 30°
Closure Type Hinged
Color White
Material Acetal Polymer
Product Type Biopsy Cassette