Learn More
Unity Lab Services™ Thermo Scientific™ Liquid Nitrogen Storage Equipment Validation Documention for Thermo Scientific™ CryoPlus™ Storage Systems
Description
IQ/OQ documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept upto date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
Specifications
Specifications
| For Use With (Equipment) | Thermo Scientific Liquid Nitrogen Storage Equipment including Thermo Scientific CryoPlus™ Storage Systems |
| Description | Documentation Only for the following, Services Not Included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, open door and power loss testing. |
| Location | Contact your local sales representative for a list of standard and extended coverage areas. |
| Includes | IQ/OQ Documentation |