Learn More
Thermo Scientific™ HPLC Syringes for CTC Instruments
HPLC syringes offer easy, accurate, and reproducible injections.
Brand: Thermo Scientific™ 365DL991
Specifications
22 G | |
HPLC Syringe | |
10 μL | |
Fixed |
1.9 in. | |
Ea. | |
CTC Instruments |
Product Content Correction
Product Title
Thermo Scientific™ HPLC Syringes for CTC Instruments > Gas tight; 10μL; 51mm; 22 gauge; Fixed needle