Learn More
Thermo Scientific™ EPD Mk2, G and N2 Readers
Shop All Thermo Scientific Products
Click to view available options
Weight (Metric):
50 g
Length (Metric) Cable:
1 m
Unit Size:
Each
Description
Compatible with the full range of Mk2 software products
- Connect directly to the PC via a serial or optional USB interface
- Require no power supply
- For more demanding environments, and where it is necessary to ensure that only one specific EPD can be “seen” by the reader, the desktop model ACT-5 may be used
- The IrDA and ACT-5 are compatible with EasyEPD2 and EasyIssue software
Specifications
Specifications
| Weight (English) | 0.1 lb. |
| Weight (Metric) | 50 g |
| Connector Types | Serial RS232 or USB 1.1 and 2.0 compatible interface direct to PC |
| For Use With (Equipment) | EasyEPD2 and EasyIssue software |
| For Use With (Application) | For office Environments and Mobile applications |
| Includes | Easy EPD software, USB interface |
| Length (English) Cable | 3.2 ft. |
| Length (Metric) Cable | 1 m |
| Product Type | Basic Irda Reader |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido