Learn More
Thermo Scientific™ CryoMed™ Thermocouple Sensors
Sample thermocouple sensors
Brand:  Thermo Scientific™ 4000381
 View more versions of this product
Product Code. 10001164
  • € 364.82 / Each
Estimated Shipment: 08-05-2025
to see stock.
Explore more special offers
Description
Sample thermocouple sensors
Thermocouple sensors for cryo tubes, sheath straw and vial samples.
  • Include male plug
For controlled-rate freezing applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thermocouple Sensor
CryoMed 7450